Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044696.1 | NP_001038161.1 | 11 | 1386 | ||
XM_006232235.2 | XP_006232297.1 | 12 | 1357 | |||
XM_008760937.1 | XP_008759159.1 | 12 | 1448 | |||
XM_008760938.1 | XP_008759160.1 | 11 | 1144 | |||
GenBank mRNA | BC097432.1 | - | 11 | 1386 | 7455342 |