Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234797.2 | XP_006234859.1 | 2 | 994 | ||
XM_006234798.2 | XP_006234860.1 | 2 | 1529 | |||
XM_006234799.2 | XP_006234861.1 | 2 | 1053 | |||
XM_006234800.2 | XP_006234862.1 | 2 | 994 | |||
XM_008762120.1 | XP_008760342.1 | 2 | 1295 |