Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFTUD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142605.1 | NP_001136077.1 | 5, 6 | 650 | ||
NM_001258353.1 | NP_001245282.1 | 6, 7 | 896 | |||
NM_001258354.1 | NP_001245283.1 | 6, 7 | 729 | |||
NM_004247.3 | NP_004238.3 | 6, 7 | 759 | |||
GenBank mRNA | AJ505017.1 | - | 2, 3 | 133 | ||
AK126464.1 | - | 7, 8 | 565 | |||
AK128148.1 | - | 6, 7 | 896 | |||
AK225335.1 | - | 6, 7 | 519 | |||
AK297392.1 | - | 6, 7 | 535 | |||
AK302828.1 | - | 6, 7 | 399 | |||
AK316098.1 | - | 5, 6 | 470 | |||
BC002360.2 | - | 6, 7 | 529 | 2960986 | ||
CR456774.1 | - | 5, 6 | 498 | |||
D21163.1 | - | 6, 7 | 558 |