Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103690020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235962.2 | XP_006236024.1 | 11 | 1756 | ||
XM_006235963.2 | XP_006236025.1 | 1329 | ||||
XM_006235964.2 | XP_006236026.1 | 11 | 1772 | |||
XM_006235965.2 | XP_006236027.1 | 11 | 1761 |
Gene Symbol: RGD1565355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109218.1 | NP_001102688.1 | 1121 | |||
GenBank mRNA | BC091104.1 | - | 1319 | 7383442 |