Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204077.1 | NP_001191006.1 | 3 | 313 | ||
NM_004788.3 | NP_004779.2 | 3 | 313 | |||
GenBank mRNA | AF087968.1 | - | 3 | 252 | ||
AK296369.1 | - | 3 | 260 | |||
AL713515.1 | - | 3 | 312 | |||
BC030973.1 | - | 3 | 262 | |||
BC052643.1 | - | 3 | 271 | |||
BC062695.1 | - | 3 | 293 | |||
BC111417.1 | - | 3 | 287 | 40038776 | ||
BC112367.1 | - | 3 | 290 | 40038767 | ||
BU167020.1 | - | 3 | 270 | |||
D50916.1 | - | 3 | 250 | |||
DB068702.1 | - | 3 | 313 |