Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cidec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024333.2 | NP_001019504.2 | 4 | 322 | ||
NM_001244797.1 | NP_001231726.1 | 4 | 334 | |||
NM_001244798.1 | NP_001231727.1 | 4 | 323 | |||
GenBank mRNA | BC083860.1 | - | 4 | 322 | 7126532 | |
FM043477.1 | - | 4 | 331 | |||
FM103335.1 | - | 4 | 323 |