Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029908.1 | NP_001025079.1 | 6 | 647 | ||
XM_008763323.1 | XP_008761545.1 | 6 | 1017 | |||
XM_008763324.1 | XP_008761546.1 | 6 | 1017 | |||
GenBank mRNA | AY197774.1 | - | 6 | 647 |
Gene Symbol: Klrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019261.1 | NP_062134.1 | 6 | 674 | ||
GenBank mRNA | AF021349.1 | - | 6 | 674 |