Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tuba3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024336.1 | NP_001019507.1 | 3 | 389 | ||
GenBank mRNA | BC079242.1 | - | 3 | 389 | 7132665 |