Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031689.2 | NP_001026859.1 | 5 | 1095 | ||
NM_001321546.1 | NP_001308475.1 | 5 | 1098 | |||
XM_011518072.2 | XP_011516374.1 | 5 | 1084 | |||
XM_017015319.1 | XP_016870808.1 | 5 | 1084 | |||
GenBank mRNA | AF083395.1 | - | 4 | 499 | ||
AF145020.1 | - | 5 | 894 | |||
AJ238243.1 | - | 4 | 527 | |||
AK001642.1 | - | 5 | 798 | |||
AK002143.1 | - | 5 | 858 | |||
AK022761.1 | - | 5 | 773 | |||
AK223544.1 | - | 4 | 530 | |||
AL133608.1 | - | 5 | 1088 | |||
BC032551.1 | - | 5 | 777 | 5555038 |