Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031563.3 | NP_113751.3 | 2 | 312 | ||
XM_008764058.1 | XP_008762280.1 | 2 | 352 | |||
XM_008764059.1 | XP_008762281.1 | 2 | 552 | |||
GenBank mRNA | AB046928.2 | - | 2 | 471 | ||
BC072486.1 | - | 2 | 312 | 7105037 | ||
BC098672.1 | - | 2 | 312 | 7124218 | ||
BC103637.1 | - | 2 | 306 | 7107271 | ||
D13309.1 | - | 2 | 282 | |||
M57299.1 | - | 2 | 288 | |||
M69138.1 | - | 2 | 448 |