Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191576.1 | NP_001178505.1 | 3 | 672 | ||
XM_006239392.1 | XP_006239454.1 | 3 | 694 | |||
XM_006239395.2 | XP_006239457.1 | 3 | 475 | |||
XM_006239396.2 | XP_006239458.2 | 3 | 429 |