Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191986.1 | NP_001178915.1 | 1243 | |||
XM_006241085.2 | XP_006241147.1 | 2395 | ||||
XM_006241094.2 | XP_006241156.1 | 312 |