Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256672.1 | NP_001243601.1 | 7 | 2012 | ||
NM_001256673.1 | NP_001243602.1 | 6 | 1865 | |||
NM_001256674.1 | NP_001243603.1 | 6 | 1862 | |||
NM_001256675.1 | NP_001243604.1 | 6 | 1802 | |||
NM_001256676.1 | NP_001243605.1 | 7 | 1826 | |||
NM_001256677.1 | NP_001243606.1 | 8 | 2143 | |||
NM_001324226.1 | NP_001311155.1 | 6 | 2131 | |||
NM_001324227.1 | NP_001311156.1 | 7 | 1829 | |||
NM_001324228.1 | NP_001311157.1 | 7 | 1893 | |||
NM_001324229.1 | NP_001311158.1 | 6 | 2320 | |||
NM_001324230.1 | NP_001311159.1 | 8 | 2146 | |||
NM_004809.4 | NP_004800.2 | 7 | 2015 | |||
XM_005254792.4 | XP_005254849.1 | 7 | 2919 | |||
XM_006720773.3 | XP_006720836.1 | 7 | 2917 | |||
GenBank mRNA | AF074953.1 | - | 7 | 1876 | ||
AF111800.1 | - | 6 | 1814 | |||
AF156557.1 | - | 7 | 2023 | |||
AK075244.1 | - | 7 | 1911 | |||
AK093689.1 | - | 3 | 2280 | |||
AK302051.1 | - | 6 | 1820 | |||
AL109664.1 | - | 6 | 1692 | |||
AL109665.1 | - | 7 | 1866 | |||
BC034379.1 | - | 7 | 1961 | 5185908 | ||
Y16522.1 | - | 7 | 1921 |