Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plscr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191999.1 | NP_001178928.1 | 3 | 331 | ||
XM_006243540.2 | XP_006243602.1 | 5 | 666 |