Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109314.1 | NP_001102784.1 | 2 | 1814 | ||
XM_006257481.2 | XP_006257543.1 | 2 | 1740 |
Gene Symbol: LOC100910807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752135.3 | XP_003752183.1 | 1913 | |||
XM_006257485.2 | XP_006257547.1 | 1740 |