Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773293.1 | XP_008771515.1 | 2 | 314 |
Gene Symbol: LOC102546630 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257041.2 | XP_006257103.1 | 2 | 378 |