Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145457.2 | NP_001138929.2 | 4 | 427 | ||
NM_001145458.2 | NP_001138930.2 | 4 | 427 | |||
NM_001242356.2 | NP_001229285.1 | 3 | 142 | |||
NM_001242357.2 | NP_001229286.1 | 3 | 142 | |||
NM_004829.6 | NP_004820.2 | 4 | 427 | |||
XM_011527528.2 | XP_011525830.1 | 4 | 535 | |||
XM_011527529.2 | XP_011525831.1 | 4 | 531 | |||
XM_011527530.2 | XP_011525832.1 | 4 | 523 | |||
GenBank mRNA | AJ001383.1 | - | 4 | 428 | ||
AJ006121.1 | - | 4 | 396 | |||
AJ006122.1 | - | 3 | 111 | |||
AJ006123.1 | - | 3 | 111 | |||
AY346373.1 | - | 2 | 68 | |||
BC064806.1 | - | 4 | 414 | 5218848 |