Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252044.2 | XP_006252106.2 | 8 | 826 | ||
XM_008770696.1 | XP_008768918.1 | 7 | 809 | |||
XM_008770697.1 | XP_008768919.1 | 8 | 877 | |||
XM_008770698.1 | XP_008768920.1 | 8 | 852 | |||
XM_008770699.1 | XP_008768921.1 | 7 | 810 | |||
XM_008770700.1 | XP_008768922.1 | 5 | 594 |