Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192009.1 | NP_001178938.1 | 7 | 843 | ||
XM_006255721.2 | XP_006255783.1 | 7 | 2553 | |||
XM_006255722.2 | XP_006255784.1 | 8 | 2201 | |||
XM_008772644.1 | XP_008770866.1 | 7 | 2553 | |||
XM_008772645.1 | XP_008770867.1 | 7 | 2553 |