Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006228205.2 | XP_006228267.1 | 8 | 2788 | ||
XM_006228206.2 | XP_006228268.1 | 7 | 1812 | |||
XM_006228207.2 | XP_006228269.1 | 6 | 1757 | |||
XM_006228208.2 | XP_006228270.1 | 7 | 2701 | |||
XM_008759006.1 | XP_008757228.1 | 7 | 1890 | |||
XM_008759007.1 | XP_008757229.1 | 7 | 1827 |