Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkr-p1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040189.2 | NP_001035279.1 | 3, 4 | 294 | ||
GenBank mRNA | AF525533.1 | - | 3, 4 | 262 | ||
DQ157010.1 | - | 3, 4 | 294 | |||
EF100678.1 | - | 3, 4 | 262 | |||
JF739552.1 | - | 3, 4 | 262 |