Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC4A7 SIRNA Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
UniGene ID:
Hs.250072
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18208
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC4A7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001258379.1
NP_001245308.1
5
831
NM_001258380.1
NP_001245309.1
5
593
NM_001321103.1
NP_001308032.1
5
843
NM_001321104.1
NP_001308033.1
5
843
NM_001321105.1
NP_001308034.1
5
843
NM_001321106.1
NP_001308035.1
5
831
NM_001321107.1
NP_001308036.1
5
831
NM_001321108.1
NP_001308037.1
5
843
NM_003615.4
NP_003606.3
5
593
NR_135541.1
5
831
NR_135542.1
5
831
NR_135543.1
5
831
NR_135544.1
5
831
XM_005265598.4
XP_005265655.1
5
1097
XM_005265600.4
XP_005265657.1
5
1098
XM_006713421.2
XP_006713484.1
5
831
XM_011534256.2
XP_011532558.1
5
1104
XM_011534258.1
XP_011532560.1
5
807
XM_011534261.2
XP_011532563.1
5
1108
XM_011534263.2
XP_011532565.1
5
807
XM_011534265.1
XP_011532567.1
5
807
XM_017007526.1
XP_016863015.1
6
728
XM_017007527.1
XP_016863016.1
5
831
GenBank mRNA
AB012130.1
-
2
279
AF047033.1
-
5
593
AF053755.1
-
2
271
AF089726.1
-
5
593
AK309734.1
-
5
831
EU499349.1
-
5
758
EU934246.1
-
5
758
EU934247.1
-
5
770
EU934248.1
-
5
770
EU934249.1
-
5
758
EU934250.1
-
5
770
FJ178574.1
-
5
758
FJ178575.1
-
5
758
FJ178576.1
-
5
758
GU354307.1
-
5
758
GU354308.1
-
5
770
GU354309.1
-
5
770
GU354310.1
-
5
770
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_058211, NM_001270861, NM_001270860, XM_008770482, XM_008770483, and others.
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0