Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253676.1 | NP_001240605.1 | 3, 4 | 822 | ||
NM_004865.3 | NP_004856.1 | 3, 4 | 524 | |||
XM_017011513.1 | XP_016867002.1 | 5, 6 | 1097 | |||
GenBank mRNA | AB020881.1 | - | 3, 4 | 439 | ||
AF130312.1 | - | 3, 4 | 467 | |||
AF136570.1 | - | 3, 4 | 471 | |||
AK292320.1 | - | 3, 4 | 777 | |||
AK313645.1 | - | 3, 4 | 492 | |||
BC000381.2 | - | 4, 5 | 492 | 2820491 | ||
BC017559.2 | - | 3, 4 | 435 | 3911352 | ||
BI560657.1 | - | 3, 4 | 529 |