Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC679894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250641.2 | XP_006250703.1 | 2 | 2376 | ||
XM_006250642.2 | XP_006250704.1 | 2 | 2376 | |||
XM_006250643.2 | XP_006250705.1 | 2 | 2376 | |||
XM_008770063.1 | XP_008768285.1 | 2 | 2376 | |||
XM_008770064.1 | XP_008768286.1 | 2 | 2376 | |||
XM_008770065.1 | XP_008768287.1 | 2 | 2376 | |||
XM_008770066.1 | XP_008768288.1 | 2 | 2376 | |||
XM_008770067.1 | XP_008768289.1 | 2 | 2376 | |||
XM_008770068.1 | XP_008768290.1 | 2 | 2376 | |||
XM_008770069.1 | XP_008768291.1 | 2 | 4380 |