Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749245.3 | XP_003749293.1 | 3 | 1034 | ||
GenBank mRNA | FQ213604.1 | - | 3 | 960 |