Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ckap2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057508.5 | XP_001057508.3 | 4 | 299 | ||
XM_006234995.2 | XP_006235057.1 | 4 | 299 |