Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127393.1 | NP_001120865.1 | 3 | 468 | ||
NM_004894.2 | NP_004885.1 | 2 | 199 | |||
GenBank mRNA | AF054175.1 | - | 2 | 162 | ||
AF116639.1 | - | 195 | ||||
AK309355.1 | - | 2 | 160 | |||
AK309499.1 | - | 2 | 162 | |||
AK312099.1 | - | 2 | 160 | |||
BC000429.2 | - | 2 | 151 | 2819801 | ||
BC001944.1 | - | 2 | 151 | 2819801 | ||
BX161388.1 | - | 3 | 403 | |||
CB127906.1 | - | 3 | 427 | |||
DA526965.1 | - | 2 | 199 |