Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001057850.5 | XP_001057850.2 | 3 | 3343 | ||
XM_006236792.2 | XP_006236854.1 | 3 | 3017 | |||
XM_006236793.2 | XP_006236855.1 | 2 | 2960 | |||
XM_006236794.2 | XP_006236856.1 | 2 | 2386 | |||
XM_008763094.1 | XP_008761316.1 | 4 | 2641 | |||
XM_008763095.1 | XP_008761317.1 | 4 | 5388 |