Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020634.1 | NP_065685.1 | 2 | 603 | ||
GenBank mRNA | AF110641.1 | - | 2 | 578 | ||
AF263538.1 | - | 2 | 603 | |||
AY358875.1 | - | 2 | 632 | |||
BC030959.1 | - | 2 | 615 | 4610141 |