Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001059906.5 | XP_001059906.3 | 10, 11 | 2178 | ||
GenBank mRNA | BC169031.1 | - | 10, 11 | 1236 |