Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127684.2 | NP_001121156.1 | 3, 4 | 303 | ||
GenBank mRNA | BC167115.1 | - | 3, 4 | 302 | 6922256 | |
FM055502.1 | - | 3, 4 | 199 | |||
FM088467.1 | - | 3, 4 | 283 | |||
FQ210865.1 | - | 3, 4 | 258 | |||
FQ217628.1 | - | 3, 4 | 246 | |||
FQ224471.1 | - | 3, 4 | 248 | |||
FQ224731.1 | - | 3, 4 | 274 |