Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109442.1 | NP_001102912.1 | 5 | 642 | ||
XM_006240290.2 | XP_006240352.1 | 4 | 344 | |||
XM_008764848.1 | XP_008763070.1 | 4 | 326 | |||
GenBank mRNA | BC166864.1 | - | 4 | 259 | 8367571 | |
FQ224144.1 | - | 4 | 349 |