Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270531.1 | NP_001257460.1 | 5, 6 | 481 | ||
NM_001270532.1 | NP_001257461.1 | 5, 6 | 613 | |||
NM_001270533.1 | NP_001257462.1 | 5, 6 | 481 | |||
NM_004906.4 | NP_004897.2 | 5, 6 | 613 | |||
NM_152857.2 | NP_690596.1 | 5, 6 | 481 | |||
NM_152858.2 | NP_690597.1 | 5, 6 | 613 | |||
XM_017011514.1 | XP_016867003.1 | 5, 6 | 451 | |||
XM_017011515.1 | XP_016867004.1 | 3, 4 | 713 | |||
XM_017011516.1 | XP_016867005.1 | 5, 6 | 451 | |||
GenBank mRNA | AF277190.1 | - | 1, 2 | 53 | ||
AF374416.1 | - | 5, 6 | 494 | |||
AJ276706.1 | - | 4, 5 | 261 | |||
AK127822.1 | - | 5, 6 | 492 | |||
AK290854.1 | - | 5, 6 | 494 | |||
BC000383.2 | - | 5, 6 | 472 | |||
BC004432.1 | - | 5, 6 | 472 | |||
BC069192.1 | - | 5, 6 | 449 | 4653831 | ||
BG702882.1 | - | 5, 6 | 485 | |||
BQ948913.1 | - | 5, 6 | 419 | |||
BX387163.2 | - | 5, 6 | 463 | |||
CN284477.1 | - | 5, 6 | 443 | |||
D14661.1 | - | 5, 6 | 393 | |||
DA561768.1 | - | 5, 6 | 487 |