Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gstz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109445.1 | NP_001102915.1 | 2, 3 | 184 | ||
XM_006240399.2 | XP_006240461.1 | 2, 3 | 158 | |||
GenBank mRNA | BC158833.1 | - | 2, 3 | 126 | 7381517 | |
FJ179410.1 | - | 1, 2 | 61 |