Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTRF1 SIRNA Assays ›
Gene Symbol:
MTRF1
Gene Name:
mitochondrial translational release factor 1
Gene Aliases:
MRF1, MTTRF1, RF1
Chromosome Location:
Chr.13: 41216369 - 41311794 on Build GRCh38
UniGene ID:
Hs.382176
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18477
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTRF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004294.2
NP_004285.2
2
222
XM_005266599.2
XP_005266656.1
4
471
XM_006719897.2
XP_006719960.1
4
579
XM_006719898.2
XP_006719961.1
4
563
XM_006719900.2
XP_006719963.1
3
292
XM_011535315.2
XP_011533617.1
4
688
XM_011535317.2
XP_011533619.1
3
571
XM_011535318.2
XP_011533620.1
4
731
XM_011535320.2
XP_011533622.1
4
667
XM_011535321.2
XP_011533623.1
7
803
XM_011535323.2
XP_011533625.1
4
533
XM_011535325.2
XP_011533627.1
3
486
XM_011535326.2
XP_011533628.1
3
532
XM_011535327.2
XP_011533629.1
3
532
XM_017020856.1
XP_016876345.1
3
532
XM_017020857.1
XP_016876346.1
6
666
XM_017020858.1
XP_016876347.1
5
661
XM_017020859.1
XP_016876348.1
3
532
XM_017020860.1
XP_016876349.1
5
549
XM_017020861.1
XP_016876350.1
5
586
XM_017020862.1
XP_016876351.1
4
1744
XM_017020863.1
XP_016876352.1
4
1592
XM_017020864.1
XP_016876353.1
4
1484
XM_017020865.1
XP_016876354.1
3
1436
XM_017020866.1
XP_016876355.1
2
1301
XM_017020867.1
XP_016876356.1
4
449
XM_017020868.1
XP_016876357.1
2
417
XM_017020869.1
XP_016876358.1
3
315
XM_017020870.1
XP_016876359.1
4
579
XM_017020871.1
XP_016876360.1
3
292
XM_017020872.1
XP_016876361.1
3
532
XM_017020873.1
XP_016876362.1
7
873
XM_017020874.1
XP_016876363.1
4
425
XM_017020875.1
XP_016876364.1
4
419
XM_017020876.1
XP_016876365.1
2
1635
XM_017020877.1
XP_016876366.1
4
470
XM_017020878.1
XP_016876367.1
4
471
XM_017020879.1
XP_016876368.1
3
1544
GenBank mRNA
AF072934.1
-
2
193
AK055086.1
-
4
593
AK294343.1
-
3
379
BC027731.2
-
3
498
BC042196.1
-
2
222
5198308
CA489782.1
-
1
532
Back To Top
More Information
Panther Classification:
Molecular Function -
translation release factor
Gene Ontology Categories:
Function(s)
Process(es)
regulation of translational termination
mitochondrial translational termination
translation release factor activity
translation release factor activity, codon specific
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-whlt2:80/10.239.162.50:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0