Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006536.5 | NP_006527.1 | 14 | 2895 | ||
GenBank mRNA | AB026833.1 | - | 14 | 2774 | ||
AF043977.1 | - | 14 | 2841 | |||
AF127980.1 | - | 14 | 2733 | |||
AK290348.1 | - | 14 | 2800 | |||
BC041096.1 | - | 14 | 2796 | 5752653 |