Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxpe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109435.1 | NP_001102905.1 | 3 | 1142 | ||
XM_006248252.2 | XP_006248314.1 | 3 | 968 | |||
XM_006248253.2 | XP_006248315.1 | 3 | 979 |