Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3PXD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014631.2 | NP_055446.2 | 8 | 642 | ||
XM_005270294.4 | XP_005270351.1 | 8 | 691 | |||
XM_005270295.4 | XP_005270352.1 | 7 | 645 | |||
XM_005270297.3 | XP_005270354.1 | 2 | 1922 | |||
XM_005270298.4 | XP_005270355.1 | 2 | 725 | |||
XM_006718079.3 | XP_006718142.1 | 8 | 695 | |||
XM_006718080.3 | XP_006718143.1 | 2 | 726 | |||
XM_011540385.2 | XP_011538687.1 | 7 | 649 | |||
XM_011540386.2 | XP_011538688.1 | 2 | 1925 | |||
XM_017016969.1 | XP_016872458.1 | 8 | 700 | |||
XM_017016970.1 | XP_016872459.1 | 6 | 432 | |||
GenBank mRNA | AB007878.1 | - | 6 | 392 | ||
AK056469.1 | - | 7 | 365 | |||
AK294454.1 | - | 2 | 98 | |||
AK294736.1 | - | 3 | 220 | |||
AK295314.1 | - | 2 | 175 | |||
AK295660.1 | - | 3 | 286 | |||
AK302015.1 | - | 5 | 314 | |||
AK304828.1 | - | 2 | 114 | |||
AK315983.1 | - | 3 | 225 | |||
CN307901.1 | - | 7 | 596 |