Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kti12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044288.1 | NP_001037753.1 | 1 | 557 | ||
GenBank mRNA | BC088196.1 | - | 1 | 557 | 7374597 |