Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdhd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109511.1 | NP_001102981.1 | 2 | 697 | ||
GenBank mRNA | BC166898.1 | - | 2 | 694 | 6922018 |