Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phemx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728846.4 | XP_002728892.3 | 1 | 2457 | ||
XM_006230915.2 | XP_006230977.1 | 1 | 2457 | |||
XM_006230916.2 | XP_006230978.1 | 1 | 2457 | |||
GenBank mRNA | BC088266.1 | - | 1 | 12 | ||
BC105909.1 | - | 1 | 73 |