Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014663.2 | NP_055478.2 | 19 | 2987 | ||
XM_005271354.3 | XP_005271411.1 | 19 | 3024 | |||
XM_005271355.3 | XP_005271412.1 | 19 | 2942 | |||
XM_005271356.3 | XP_005271413.1 | 20 | 3063 | |||
XM_011542467.2 | XP_011540769.2 | 10 | 2095 | |||
XM_017002912.1 | XP_016858401.1 | 20 | 3132 | |||
GenBank mRNA | AB014577.1 | - | 19 | 2946 | ||
AK294596.1 | - | 5 | 572 | |||
AK310295.1 | - | 4 | 605 | |||
BC002558.2 | - | 19 | 2955 | 3138875 |