Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232102.2 | XP_006232164.1 | 3 | 486 | ||
XM_006232103.2 | XP_006232165.1 | 3 | 755 | |||
XM_006232104.2 | XP_006232166.1 | 3 | 210 | |||
XM_006232105.2 | XP_006232167.1 | 3 | 510 | |||
XM_006232106.2 | XP_006232168.1 | 3 | 218 | |||
XM_008760847.1 | XP_008759069.1 | 3 | 798 |