Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014671.2 | NP_055486.2 | 23 | 3507 | ||
XM_005249564.4 | XP_005249621.1 | 22 | 3441 | |||
XM_017012818.1 | XP_016868307.1 | 24 | 3779 | |||
GenBank mRNA | AK127280.1 | - | 8 | 3778 | ||
AK295138.1 | - | 23 | 3187 | |||
D13635.1 | - | 23 | 3498 |