Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_012288.3 | NP_036420.1 | 7, 8 | 770 | ||
XM_011515005.2 | XP_011513307.1 | 7, 8 | 645 | |||
GenBank mRNA | AK291751.1 | - | 7, 8 | 656 | ||
AK311560.1 | - | 7, 8 | 639 | |||
AY927571.1 | - | LIT3572 AY927571 |
1, 2 | 336 | ||
BC028121.1 | - | 7, 8 | 770 | 5169808 | ||
D31762.1 | - | 7, 8 | 696 | |||
KU178488.1 | - | 7, 8 | 621 | |||
KU178489.1 | - | 6, 7 | 557 |