Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qki | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758777.1 | XP_008756999.1 | 4, 5 | 1015 | ||
XM_008758779.1 | XP_008757001.1 | 1015 | ||||
XM_008758780.1 | XP_008757002.1 | 1014 |
Gene Symbol: Qk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115021.1 | NP_001108493.1 | 6, 7 | 875 | ||
GenBank mRNA | AB054997.1 | - | 3, 4 | 353 | ||
BC158800.1 | - | 6, 7 | 875 | 7320367 |