Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014680.3 | NP_055495.2 | 17, 18 | 3360 | ||
XM_005258073.2 | XP_005258130.1 | 17, 18 | 3355 | |||
XM_017025423.1 | XP_016880912.1 | 16, 17 | 3209 | |||
GenBank mRNA | BC120873.1 | - | 17, 18 | 3219 | 40075102 | |
CR933634.1 | - | 18, 19 | 3222 | |||
D43947.3 | - | 17, 18 | 3332 |