Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010989.2 | NP_001010989.1 | 6 | 955 | ||
NM_014685.3 | NP_055500.1 | 6 | 958 | |||
GenBank mRNA | AB034989.1 | - | 6 | 901 | ||
AF055001.1 | - | 6 | 898 | |||
AF055003.1 | - | 6 | 698 | |||
AF217990.1 | - | 3 | 1239 | |||
AK223233.1 | - | 6 | 901 | |||
AK291565.1 | - | 6 | 901 | |||
AK292088.1 | - | 6 | 901 | |||
AK302167.1 | - | 5 | 862 | |||
BC000086.1 | - | 6 | 893 | 3504515 | ||
BC008320.1 | - | 6 | 892 | 3350209 | ||
BC032673.1 | - | 6 | 900 | 5575914 | ||
CR457116.1 | - | 6 | 803 | |||
D14695.1 | - | 6 | 896 |