Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010989.2 | NP_001010989.1 | 8 | 1176 | ||
NM_001272103.1 | NP_001259032.1 | 8 | 1022 | |||
NM_014685.3 | NP_055500.1 | 8 | 1179 | |||
GenBank mRNA | AB034989.1 | - | 8 | 1122 | ||
AF055001.1 | - | 8 | 1119 | |||
AF055003.1 | - | 8 | 919 | |||
AF217990.1 | - | 5 | 1460 | |||
AK223233.1 | - | 8 | 1122 | |||
AK291565.1 | - | 8 | 1122 | |||
AK292088.1 | - | 8 | 1122 | |||
AK302167.1 | - | 7 | 1083 | |||
BC000086.1 | - | 8 | 1114 | 3504515 | ||
BC008320.1 | - | 8 | 1113 | 3350209 | ||
BC009739.1 | - | 6 | 645 | 4095997 | ||
BC032673.1 | - | 8 | 1121 | 5575914 | ||
CR457116.1 | - | 8 | 1024 | |||
D14695.1 | - | 8 | 1117 | |||
DQ837586.1 | - | 8 | 964 |