Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP6NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080491.3 | NP_001073960.1 | 14 | 1490 | ||
NM_014688.3 | NP_055503.1 | 15 | 1821 | |||
XM_006717542.3 | XP_006717605.1 | 16 | 1647 | |||
XM_011519762.2 | XP_011518064.1 | 17 | 1980 | |||
XM_011519763.2 | XP_011518065.1 | 10 | 1981 | |||
XM_017016971.1 | XP_016872460.1 | 16 | 1654 | |||
XM_017016972.1 | XP_016872461.1 | 16 | 1792 | |||
XM_017016973.1 | XP_016872462.1 | 14 | 4817 | |||
GenBank mRNA | AB449916.1 | - | 13 | 1391 | ||
AK292944.1 | - | 15 | 1611 | |||
BC042943.2 | - | 15 | 1659 | 5296060 | ||
D13644.1 | - | 15 | 1670 |